Separating funnels, Squibb, [ST] glass stopcock and stopper, KIMAX®, KIMBLE®
供应商: DWK Life Sciences
KIMAX®
KIML29048-250EA 0 CNY
KIML29048-125 KIML29048-250 KIML29048-500
Separating funnels, Squibb, [ST] glass stopcock and stopper, KIMAX®, KIMBLE®
漏斗 分液漏斗
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Very good temperature resistance
  • Very good chemical resistance
KIMAX® separating funnels with a [ST] ground glass stopcock and stopper.
Funnels come with lower stems having large internal diameter that allow complete separation and eliminates the necessity of emptying the funnel.
Certifications: Designed from ASTM Specification E1096, Type IV requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements and then RAY-SORB® processed.