Alkaline cleaning solution, liquid concentrate, Micro-90®
供应商: International Products
MICRO-90®
IPROM-9051-04EA 8425 CNY
IPROM-9033 IPROM-9051-04 IPROM-9050-12
Alkaline cleaning solution, liquid concentrate, Micro-90®
清洗剂 洗涤剂和表面清洁剂
Micro-90® is a mild, yet powerful, multipurpose, alkaline cleaning concentrate that has long been used in laboratories, industrial applications, and critical cleaning processes.
  • Manufactured in a dedicated system
  • pH: 9,5
Micro-90® is a unique chelating detergent that contains ionic and non-ionic ingredients, which combine to produce a variety of cleaning actions. It lifts, disperses, emulsifies, sequesters, suspends, and decomposes soils, then rinses away leaving the surface absolutely clean. Excellent for use on stainless steel, glass, ceramic, plastic, rubber, concrete, filter membranes and most other hard surfaces.
Certifications: Registered with NSF as an A1 cleaner and can be validated in FDA processes.