Liquid nitrogen refrigerators, LS series
供应商: Jencons
JENC232-078EA 0 CNY
JENC232-078 JENC232-028
Liquid nitrogen refrigerators, LS series
The LS series refrigerators are uniquely designed for large vial capacity in convenient box-type storage racks together with exceptionally low evaporation rates. To comply with Medical Device Directive (MDD) CE 93 / 42 Class IIa Taylor Wharton has introduced the new CryoCE range to the LS series. All MDD models feature the M505CE controller which provides automatic filling and alarm features. For details, see the information for accessories in the K series cryostorage systems catalogue entry.
  • LS750 holds 25 vials in 5×5 matrix boxes
  • LS4800, holds vials in 10×10 matrix boxes
  • Each refrigerator holds six racks with system index location ring and internal spider allows easy location and quick retrieval of samples
  • For sample protection low liquid level alarms are available on all units
  • 3 year warranty on vacuum
订购须知 The LS3000 and LS6000 are also available with controllers which provide automatic filling and alarm features. Controllers are a factory fitted option. Retrofitting is not possible. Please contact your local VWR sales office for details.
Delivery information: All units are supplied complete with racks, boxes and neck core.
Caution: ** 1,2 or 2,0 ml vial size with Ø 12,5 mm (internal thread).