J.T.Baker® BAKER INSTRA-ANALYZED®, Calcium Standard Solution, 1000 µg/L Ca, Reagent for AAS
供应商: Avantor
BAKR6448-04EA 2289 CNY
BAKR6448-04
J.T.Baker® BAKER INSTRA-ANALYZED®, Calcium Standard Solution, 1000 µg/L Ca, Reagent for AAS
Calcium standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.
Certifications: Traceable to NIST and SRM numbers.