Beakers, low form, griffin, graduated, PP, Azlon®
供应商: SciLabware
Azlon®
AZLOBDC242PEA 1448.28 CNY
AZLOBDC242P AZLOBDC240P AZLOBDC212P AZLOBDC236P AZLOBDC218P AZLOBDC224P AZLOBDC234P AZLOBDC238P
Beakers, low form, griffin, graduated, PP, Azlon®
烧杯
Griffin style beakers made of PP.
  • Good chemical resistance
  • Very good temperature resistance
  • Graduations are clearly printed in indelible solvent resistant ink
Ideal for general laboratory use.
Certifications: Complies with ISO 7056.