Manganese standard solution, 1,000 µg/L Mn, BAKER INSTRA-ANALYZED reagent for AAS, J.T. Baker®
供应商: Avantor®
J.T.Baker®
BAKR6458-04EA 1630 CNY
BAKR6458-04
Manganese standard solution, 1,000 µg/L Mn, BAKER INSTRA-ANALYZED reagent for AAS, J.T. Baker®
标准品 Cationic Standards Manganese Standards
Manganese standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.
Certifications: Traceable to NIST and SRM numbers.