Nitrogen test, TNT plus®
供应商: Hach
HACHTNT828EA 810 CNY
HACHTNT828 HACHTNT827 HACHTNT826
Nitrogen test, TNT plus®
环境试验箱
These 'Test N Tube' are used for the determination of nitrogen by the persulfate digestion method.
  • Test 'N Tube products provide safe, convenient testing
  • TNT plus vials reduce errors with bar coded vials and RFID
  • TNT plus vials require no reagent blank