Nitrate TNTplus® kit
供应商: Hach
HACHTNT836EA 760.5 CNY
HACHTNT836
Nitrate TNTplus® kit
环境试验箱
This kit is used for the determination of nitrate nitrogen by the dimethylphenol method using TNTplus vials.
  • Reduce errors with bar coded vials and RFID
  • Innovative vial
  • No reagent blank necessary