Sulphate TNTplus® kit
供应商: Hach
HACHTNT864EA 0 CNY
HACHTNT864 HACHTNT865
Sulphate TNTplus® kit
环境试验箱
This kit is used for the the determination of sulphate by the turbidimetric method using TNTplus vials.
  • Reduce errors with bar coded vials and RFID
  • Innovative vials with DosiCaps
  • No reagent blank necessary