Cadmium TNTplus® kit
供应商: Hach
HACHTNT852EA 1250 CNY
HACHTNT852
Cadmium TNTplus® kit
环境试验箱
This kit is used for the determination of cadmium by the cadion method using TNTplus™ vials.
  • Reduce errors with bar coded vials and RFID
  • Innovative vial
  • No reagent blank necessary