Alkalinity TNTplus™ kit (total)
供应商: Hach
HACHTNT870EA 690.3 CNY
HACHTNT870
Alkalinity TNTplus™ kit (total)
环境试验箱
This kit is used for the determination of alkalinity (total) by the colorimeteric method using TNTplus vials.
  • Easy and safe to handle
  • No reagent blank necessary
  • Automatic method detection