Aluminium TNTplus™ kit
供应商: Hach
HACHTNT848EA 1316.25 CNY
HACHTNT848
Aluminium TNTplus™ kit
This kit is used for the determination of aluminium by the chromazurol S method using TNTplus™ vials.
  • Easy and safe handling
  • No reagent blank necessary
  • Automatic method detection