Laboratory and scientific brushes for glassware
供应商: Perfex
PERF3642EA 0 CNY
PERF3642 PERF3652 PERF3641 PERF3651 PERF3602 PERF3624 PERF3601 PERF3600 PERF3618 PERF3617 PERF3616
Laboratory and scientific brushes for glassware
刷子
These brushes remove infectious or toxic residue from laboratory equipment.
  • Made of high-quality polyester fibres
  • Brushes will not absorb bacteria
  • Non- abrasive and odour-free
  • Compatible with gamma and ETO
  • Autoclavable at 121 °C for 30 minutes
The brushes are resistant to hot water, strong acids, petroleum distillates, benzene aromatic hydrocarbons, acetones, ketones, ethyl acetate esters and trichloroethylene hydrocarbons.
These brushes rinse clean and dry quickly. Unlike galvanised wire, stainless steel twisted wire will not rust or corrode.