DNA ladder, 100 bp
供应商: Promega
PROMG2101EA 0 CNY
PROMG2101
DNA ladder, 100 bp
电泳试剂 分子量标记
The 100 bp DNA ladder has eleven fragments that range in size from 100 to 1000 bp in 100 bp increments with an additional band at 1500 bp. The 500 bp fragment is present at increased intensity for easy identification. The ladder is dephosphorylated and ready for 5' end-labelling with radioisotopes using T4 Polynucleotide Kinase for visualisation by autoradiography.
  • Supplied at a concentration of 0,13 µg/μl
DNA Ladders have defined sizes, and are not intended for use in quantitative analysis.
Delivery information: DNA ladder is supplied with 6X blue/orange loading dye.