HPLC columns, Vydac® Denali® silica
供应商: Avantor®
Vydac Denali
HICH5174063EA 11065 CNY
HICH5174063 HICH5174054 HICH5174058 HICH5174066 HICH5174057 HICH5174061
HPLC columns, Vydac® Denali® silica
色谱柱
Vydac® Denali® (238DE) is a 120 Å C18 bonded phase with high carbon coverage, suitable for the analysis of both acidic and basic analytes. It has applications for small molecule analyses of interest to pharmaceutical and environmental laboratories.
  • High retentivity
  • LC-MS of small molecules
  • Fully scalable from capillary to process