Avantor® Prevail Amino, HPLC Columns
供应商: VWR
Avantor® Prevail
HICH99290/NEA 2555 CNY
HICH99290/N HICH99263 HICH99265
Avantor® Prevail Amino, HPLC Columns
色谱柱
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
  • Stable from highly organic to highly aqueous
  • Speciality phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
These columns can be used for carbohydrates or as a weak anion exchanger.
Accessories information: Use guard cartridges with holder (80101/N) and column coupler (HI-081).