Fluoride reagent set, CA610
供应商: Hach
HACH2812811EA 530 CNY
HACH2812811 HACH2816900
Fluoride reagent set, CA610
环境试验箱
Fluoride reagent set is used for the determination of accurate and cost-effective fluoride analysis.
Delivery information: CA610 fluoride analyser reagent set includes reagent 1 (TISAB), Standard 1 (0,5 mg/L) F, Standard 2 (5,0) mg/L F.