Portable spectrophotometer, DR2800
供应商: Hach
HACHDR2800-01B1EA 95179.5 CNY
HACHDR2800-01B1
Portable spectrophotometer, DR2800
Portable spectrophotometer for more than 240 analytical methods including more than 30 vial tests, that provide innovative bar code labelling for reliable, automatic method detection suitable for drinking and waste water.
  • USB memory stick to update the instrument or to transfer data
  • Small footprint and large touch screen interface
  • Runs on either line power or battery power
  • Accommodates multiple cell sizes and sample delivery methods
TNTplus reagent vials for selected analytical methods provide automatic method detection.
While rotating the TNT vial, the instrument takes 10 absorbance measurements in less than five seconds. The average value is used to calculate the results, providing built-in accuracy.
For a list of popular test kits for the DR2800 instrument, please see the COMPLEMENTARY PRODUCTS Tab. For a more comprehensive selection of test kits available, please search the VWR website using the search term ETK.
订购须知 An optional pour-through cell set for rapid liquid methods when high throughput analysis is needed must be ordered separately.
Delivery information: Includes printed manual, CD manual, power supply with exchangeable plug adapters (EU, GB, US, China), adapters A, B, and C with protective cover, dust cover, 1" square matched glass sample cells and lithium battery.