Monochloramine TNT reagent set
供应商: Hach
HACH2805145EA 0 CNY
HACH2805145
Monochloramine TNT reagent set
环境试验箱
For determination of Monochloramine by the Indophenol Test 'N Tube™ method.
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering
Up to 50 tests/reagent set.