Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

供应商: Promega
PROMW4021EA 0 CNY
PROMW4021
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
抗体
Ideal for use in Western Blot, ELISA and Dot Blot.

Recommended Dilutions: ELISA/Western Blot: 1:2500

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR