Thermolysin
供应商: Promega
PROMV4001EA 0 CNY
PROMV4001
Thermolysin
Thermolysin is a thermostable metalloproteinase. The high digestion temperature stability of this enzyme may be useful as an alternative to denaturants to improve digestion of proteolytically resistant proteins.
  • Optimal digestion temperature is 65 to 85 °C
  • Optimal activity at pH 5,0 to 8,5
  • Digestion of proteolytically resistant proteins
It preferentially cleaves at the N-terminus of the hydrophobic residues leucine, phenylalanine, valine, isoleucine, alanine and methionine.
Applications:
Peptide mapping.
Protein identification.
Characterisation of post-translational modifications.