VWR®, Water Baths, Digital
供应商: VWR
VWR®
VWRU89501-478EA 28593.48 CNY
VWRU89501-478 VWRU89501-466 VWRU89501-474 VWRU89501-462 VWRU89501-470_D VWRU89501-476 VWRU89501-464 VWRU89501-470 VWRU89501-472
VWR®, Water Baths, Digital
浴锅 非搅拌水浴锅
These digital general purpose water baths are sleek and cleanly designed to provide absolute control, precision and convenience.
  • Displays actual and set point temperature simultaneously
  • Three push-button pre-sets for frequently used temperatures
  • Built-in countdown timer
  • Stainless steel 304 reservoirs
Easy to use digital temperature controller features three temperature pre-sets for one-touch selection of frequently used set-points plus a user-settable high limit alarm that alerts you if the bath temperature exceeds pre-set limits.
Delivery information: Supplied with lid and removable bottom diffuser tray.