Cleaning brushes for test tubes, tufted end
供应商: Mill-Rose
MILM72034WEA 219.77 CNY
MILM72033W MILM72034W
Cleaning brushes for test tubes, tufted end
刷子
These brushes can be used to clean test tubes and narrow-mouth bottles.
  • Feature white natural bristles with black nylon tuft
  • A twisted wire handle with a loop is provided for hanging