pH buffer pouches, NIST, SINGLET™
供应商: VWR Chemicals
SINGLET™
VWRU89236-552EA 1850 CNY
VWRU89236-552 VWRU89236-550 VWRU89236-548
pH buffer pouches, NIST, SINGLET™
缓冲(液) 酸碱缓冲液
These pH buffers used for sympHony® pH meter standardisation available in sealed foil pouches.
  • Ideal for on-site analyses
  • Easy to use
  • Fresh and reliable