Bottles, wide neck, amber, protect, with GL 80® thread, DURAN®
供应商: DWK Life Sciences
SCOT218665432EA 0 CNY
SCOT218665432 SCOT218666334 SCOT218667339 SCOT218664436
Bottles, wide neck, amber, protect, with GL 80® thread, DURAN®
玻璃瓶
DURAN®, borosilicate glass 3.3, Protect, amber, coated, without screw caps and pouring rings.
  • Scratch, leak and shatter resistant, highly transparent
  • Temperature resistance from −30 to +135 °C
  • Suitable for use in microwaves
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Ideally for transport and storage of hazardous media or valuable samples. UV protection up to approx. 380 nm wavelength.