VWR®Spinbar®, Magnetic stirring bars, disposable
供应商: Avantor
VWR® Spinbar® VWR®
VWRU58947-146EA 5434.41 CNY
VWRU58947-146 VWRU58947-144 VWRU58947-142 VWRU58947-140
VWR®Spinbar®, Magnetic stirring bars, disposable
搅拌器 磁力搅拌子
The multifaceted stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Ideal for single use applications such as mixing radioactive reagents
  • Disposable; eliminates potential for cross-contamination
  • Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert
For single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.