They are general-purpose, upright laboratory refrigerators features a microprocessor temperature controller with digital display that allows precise temperature adjustment, maintenance and recovery.
- User-adjustable alarms
- Extensive security package
- Precise temperature adjustment
Units also feature an extensive temperature security package, including user-adjustable audible and visual alarms for high and low temperatures and power failures, battery backup, a temporary muting option with intermittent chirp reminder, sample simulated digital temperature display, and sensor error warning. Remote alarm contacts with adjustable delay simplify connection to central alarm systems and phone dialers. A solution bottle is installed within each unit to provide an accurate indication of sample temperatures. Refrigerators are cycle preset to maintain an average temperature of 4 °C.
Certifications: UL and C-UL listed.
Delivery information: Supplied with swivel casters (102 mm), shelves, two-year parts and labor warranty, with an additional three-year warranty on compressor parts.