VWR®, Conductivity standards, NaCl
供应商: Avantor
VWRU89236-546EA 1125 CNY
VWRU89236-546 VWRU89236-544 VWRU89236-540
VWR®, Conductivity standards, NaCl
标准品 电导率标准
NaCl standard solutions can be used to calibrate instruments that measure any samples containing NaCl (sodium chloride) as major component, such as seawater, brackish water, etc.
  • Used for calibration and accuracy checks in conductivity measurements