VWR®, General Purpose Refrigerator/Freezer
供应商: VWR
VWR®
VWRU97058-842EA 43956 CNY
VWRU97058-842 VWRU97058-840 VWRU89498-682 VWRU97055-710
VWR®, General Purpose Refrigerator/Freezer
冰箱
These are general-purpose, upright laboratory refrigerator/freezers which feature a mechanical dial thermostat, leveling legs, integrated handles, interior lights, probe access port, heavy-duty reinforced handle and a magnetic door gasket for a tight seal.
A spacious capacity allows for maximum storage space and flexibility for all types of materials. Unit operates with CFC–free refrigerant, foam insulation, and a mechanical thermostat.
Delivery information: Refrigerators and Freezers are supplied with a two-year parts and labor warranty, with an additional three-year warranty on compressor parts.