VWR® Pipette stand
供应商: VWR
VWR®
VWRU82024-540EA 944.49 CNY
VWRU82024-540 VWRU82024-542
VWR® Pipette stand
吸管架
This clear acrylic pipette stand is designed to hold most major brands of pipettes.
  • Holds three or four pipettes
  • Upper slots are 25 mm (1") wide and lower slots are 20 mm (¹³/₁₆") wide