VWR®, Wash Bottles
供应商: VWR
VWR®
VWRU414004-227EA 120.19 CNY
VWRU414004-228 VWRU414004-226 VWRU414004-227 VWRU414004-225
VWR®, Wash Bottles
洗瓶
These LDPE wash bottle features a PP screw cap and stem that are molded into a one-piece unit for leakproof operation.
Dispensing tube produces a fine stream, but can be cut for a coarser flow.