Tube racks, for colour comparison tests, SP Bel-Art
供应商: Bel-Art
Scienceware®
BELAF189160000EA 165 CNY
BELAF189160000
Tube racks, for colour comparison tests, SP Bel-Art
架子 试验管架
Colours stand out against the white surfaces of this polypropylene rack making color comparison tests easier to read.
  • Useful for drying or storing test tubes
  • Holds six (25 mm) or eight (18 mm) tubes; includes six drying pins
  • Steam autoclavable at 121 ºC