Vaccum Ovens, digital
供应商: VWR Collection
VWR®
VWRU97027-674EA 167125 CNY
VWRU97027-664 VWRU97027-674
Vaccum Ovens, digital
干燥箱 真空炉
These vacuum ovens are built with a stainless steel chamber for exceptional durability.
  • Fully programmable Watlow temperature control
  • Capable of 40-step ramp and soak profiles or 4 files With 10 steps per file
  • Digital vacuum gauge
  • RS485 communication
The doors on these units have positive latch handles with spring-loaded glass to facilitate a good vacuum seal without hinge binds that shorten the gasket life. Versatile based on the selection of gaskets and a small benchtop footprint.
The unique cross-flow ventilation forces inert gas to fill the entire chamber. To acheive required vacuum levels, can choose from a 3/8" orifice or a KF25 fitting to withstand heavy use and minimize draw-down time. Maximum permitted end vacuum is 10 μm. Leak rate is 10 μm per hour.