VWR® Clinical 200 Large Capacity Centrifuge, Bench
供应商: Avantor
VWR®
VWRU82013-814EA 47279.55 CNY
VWRU82013-814 VWRU82017-656
VWR® Clinical 200 Large Capacity Centrifuge, Bench
离心机 台式离心机
Designed for ease of use, this large-capacity centrifuge is ideal for clinical laboratory and research environments.
  • Microprocessor control
  • Safety lock lid
  • Dynamic braking with non disruptive soft stop
  • Centrifuge can be used in a cold room
  • Versatile: Accepts tubes from 1,5 ml to 50 ml, including all common blood draw tubes
A combination of digital displays and easy-set knobs allow for quick and precise setting of operating parameters. Precise operational control is maintained by the microprocessor. An imbalance sensor is incorporated into control the loop to stop operation of centrifuge in case rotor is loaded improperly. At end of run, dynamic braking brings rotor to quick and turbulence-free stop.
Delivery information: Sets include a 28° fixed-angle rotor that holds twelve 15 ml tubes and adapters for 5 ml, 7 ml, and 10 - 12 ml tubes.