VWR®, Screw Thread Vials, 8-425
供应商: VWR
VWR®
VWRU66009-878EA 1204.49 CNY
VWRU66009-878 VWRU46610-736
VWR®, Screw Thread Vials, 8-425
小瓶
Standard opening screw-thread vials are manufactured from borosilicate glass and have a maximum fill volume of 1,8 ml.
  • Screw caps with preattached septa
Closures are black and have preattached septa.