Carboys, HDPE, with versatile caps
供应商: VWR Collection
VWR®
VWRU89170-774EA 2648.89 CNY
VWRU89170-770 VWRU89170-774 VWRU89170-768
Carboys, HDPE, with versatile caps
carboy水瓶
These carboys are designed to provide users with a highly versatile container that maximises storage efficiency and ease of use.
  • Chemical-resistant
  • Cap size of 80 mm
  • Molded grips for easy lifting, handling, and pouring
  • Molded from a translucent USP Class VI, and FDA grade HDPE resin
The distinctive rectangular shape saves valuable bench space. Large handles on top of the container and molded handles in the base provide a full, secure grip to make lifting, carrying, and pouring easy. All carboys have their material and volume clearly imprinted on the outside of the bottle, as well as large, easy to read metric graduation marks which are certified to ±5% accuracy.
They offer excellent chemical resistance, making them ideal for most buffers and reagents as well as powders and solids. Open caps (sold separately) are compatible with interchangeable adapters, which feature a wide variety of top-connections, including molded-in hose barbs and quick-connect hose barbs.