VWR® General-Purpose Upright Freezers are available with manual or automatic defrost, and are designed to provide dependable, energy-efficient performance in non-critical industrial, clinical, or laboratory applications.
- Two-year parts and labor warranty
- Three-year compressor parts warranty
These freezers are manufactured with CFC-free high-density urethane foam cabinet and door insulation to maintain temperature stability. They feature a single right-hinged solid swing door, white textured steel cabinets and doors, and a magnetic door gasket to ensure a positive seal. Doors are equipped with a heavy-duty handle and keyed door lock for added security. Additional standard features include mechanical thermostat and leveling legs.
Delivery information: Supplied with door bins.