Sealing film for PCR plates
VWRU82018-844EA 1560 CNY
VWRU82018-844
Sealing film for PCR plates
This clear, polypropylene film matches the SBS standard to accurately fit PCR plates.
  • Functional temperature range is –40 to 125 °C
  • Enables accurate thermocycling
  • Designed for use with 96-well plates only
This 4 mm sealing film uses a special adhesive that works at room temperature to seal plates for accurate thermocycling and reduced evaporation without freezing or autoclaving.