Infrared thermometer
供应商: VWR, part of Avantor
VWR®
VWRU29960-032EA 4125 CNY
VWRU29960-032 VWRU89030-412
Infrared thermometer
温度计 数字温度计
Used in many laboratory and plant environments, these infrared (IR) thermometers provide non-contact °F or °C (0.1 resolution) temperature measurement of any liquid, solid, or semi-solid.
  • Simple operation
  • Backlit readout
  • Fixed emissivity of 0,95
  • Suitable for use in high temperature range applications when the user needs to be a safe distance from the target being measured
Both models feature max./min. memory, high/low alert, laser sighting, lock function for continuous measurement, average and differential, backlit readout, and auto shut off.
The –60/500°C thermometer measures both non–contact and contact temperatures with adjustable emissivity and has an 11:1 DTS. It includes a built-in thermocouple port that can accommodate any type K thermocouple with a mini-connector when direct contact measurement is required (thermocouple sold separately).
The –20/537°C thermometer offers a 12:1 DTS, a 12-dot outline laser sighting, and it is easy to store (strap and carrying case included).
Certifications: CE marked and RoHS compliant.
Delivery information: Supplied with carrying case.