These glass door laboratory refrigerators are purpose-built for scientific and medical applications.
- Adjustable operating temperature range: 1 to 10°C
- Sample simulated digital temperature display
- Audible and visual high/low temperature alarms
- Swivel casters add four inches to height
It features a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings.
Remote alarm dry contacts and an access port are provided to simplify connections to existing facility monitoring systems.
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates, as well as UL, ASHRAE compliance guidelines.
Certifications: UL/C-UL Listed.
Delivery information: Units are supplied with a two-years parts and labour warranty, with an additional five-years warranty on compressor parts.