Air flow meters with durable, one-piece clear acrylic construction and featuring a stable and easy-to-read float; threaded brass inserts for quick installation; and English or metric scales.
- Stainless steel fittings, valves, and seals
- Clear acrylic construction
- Easy disassembly and assembly for maintenance
- Maximum temperature: 65 °C
Suitable for use for applications involving air sampling equipment, gas analysers, medical systems, photoprocessing equipment, etc.
Delivery information: Supplied with stainless steel or PVC FNPT fittings, valve, and seals. Stainless steel fittings include Viton® seals. PVC fittings include Buna-N seals.