VWR®, Air Flow Meters
供应商: Avantor
VWR®
VWRU97004-732EA 3729 CNY
VWRU97004-732 VWRU97004-644 VWRU97004-696 VWRU97004-636 VWRU97004-964
VWR®, Air Flow Meters
流量计
Air flow meters with durable, one-piece clear acrylic construction and featuring a stable and easy-to-read float; threaded brass inserts for quick installation; and English or metric scales.
  • Stainless steel fittings, valves, and seals
  • Clear acrylic construction
  • Easy disassembly and assembly for maintenance
  • Maximum temperature: 65 °C
Suitable for use for applications involving air sampling equipment, gas analysers, medical systems, photoprocessing equipment, etc.
Delivery information: Supplied with stainless steel or PVC FNPT fittings, valve, and seals. Stainless steel fittings include Viton® seals. PVC fittings include Buna-N seals.