VWR®symphony™, Chromatography refrigerators with glass doors
供应商: Avantor
VWR®
VWRU97024-994EA 201511.48 CNY
VWRU97024-994 VWRU89498-678
VWR®symphony™, Chromatography refrigerators with glass doors
冰箱
These refrigerators are ideally suited for chromatography procedures requiring interior access to electrical service.
  • Microprocessor temperature control
  • Digital display
  • Adjustable alarms
Each unit is equipped with two 50 mm diameter sidewall access ports with self-closing, gasketed metal covers and a rear wall-mounted GFI protected duplex electrical outlet. A microprocessor temperature controller with digital display allows precise temperature adjustment, maintenance, and recovery. Units also feature an extensive temperature security package, including user-adjustable audible and visual alarms for high and low temperatures and power failures, battery backup, a temporary muting option with intermittent chirp reminder, sample simulated digital temperature display, and sensor error warning. Remote alarm contacts with adjustable delay simplify connection to central alarm systems and phone dialers. A solution bottle is installed within each unit to provide an accurate indication of sample temperatures. Refrigerators are cycle preset to maintain an average temperature of 4 °C.
Two-year parts and labor warranty, with an additional three-year warranty on compressor parts.
Delivery information: Supplied with 102 mm swivel casters. Shelves included.