VWR®symphony™, Flammable storage refrigerators and freezers
供应商: VWR
VWR®
VWRU97055-748EA 91717.35 CNY
VWRU89498-642 VWRU97055-740 VWRU89498-670 VWRU97055-748 VWRU89498-646
VWR®symphony™, Flammable storage refrigerators and freezers
冰箱
These storage refrigerators facilitate the storage of flammable materials, sources of sparks and ignition - such as lights, thermostats, switches and fans - have been removed from the interior of these units.
  • Exterior adjustable mechanical thermostats
  • White baked-on powder coat exterior
  • Single solid door, with keyed locks
Refrigerators are supplied with a two-year parts and labor warranty, with an additional three-year warranty on compressor parts.
Certifications: Units meet NFPA and OSHA guidelines, and are UL and C-UL listed.