Standard series manual defrost laboratory chest freezers
供应商: VWR Collection
VWR®
VWRU10819-436EA 31525 CNY
VWRU10819-436
Standard series manual defrost laboratory chest freezers
冰箱 实验室通用冰箱
These standard series manual defrost laboratory chest freezers are designed specifically with general-purpose applications in mind.
  • Adjustable operating temperature range: –15 to –25 °C
  • Application-specific compressors
  • Cold wall design
  • Supplied with two-year parts and labour warranty, and five years warranty on compressor parts
These units are ideal when stabile temperatures averaging –20 °C are required, but precise temperature control, maintenance, and alarms are not necessary. These units feature a manual defrost design to eliminate damaging temperature spikes from defrost cycles. This ensures valuable sample security. Defrosting of the unit must be performed periodically to prevent severe ice build-up.
Certifications: UL/C-UL listed.