Orbital mini shakers, VWR®
供应商: VWR, part of Avantor
VWR®
VWRU12620-938EA 15525 CNY
VWRU12620-938 VWRU10027-116
Orbital mini shakers, VWR®
振荡器 轨道振荡器
Variable-speed, microprocessor control provides consistent, uniform mixing action.
  • Digital display for speed and time
  • Microprocessor control
  • Timer with audible alarm
Ramping feature slowly increases speed to set point for added safety. Independent LED display features speed and time. Timer will display elapsed time, or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero. Ideal for use in incubators, CO2 incubators, or cold rooms. Durable base is constructed of cast aluminum for extra stability. The triple eccentric drive features permanently lubricated ball bearings. The maintenance-free, brushless DC motor provides a long service life.
Included tray can be used as a platform for mounting Erlenmeyer flask clamps, test tube racks, or securing microplates with the universal harness (sold separately). Tray features a nonskid rubber mat appropriate for holding Petri dishes and cell culture flasks. Optional universal platform with adjustable rollers holds various vessel types such as volumetric flasks or separatory funnels. The rollers open to 216 mm deep. Universal harness (sold separately) features four elastic bands and is suitable for holding Petri dishes and microplates.
Certifications: RoHS certified. CSA listed. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Delivery information: Unit includes a nonskid mat for tray and a 2,3 m detachable, three-wire cord and plug. Contact your Avantor sales representative for more information.