VWR®sympHony™, Conductivity meters, bench
供应商: VWR
VWR® symphony™
VWRU89231-678EA 21348.36 CNY
VWRU89231-676 VWRU89231-678
VWR®sympHony™, Conductivity meters, bench
电化学仪器 电导率仪
These bench top meters are specially designed for use in environmental field applications and in the laboratory. The meters are rugged and durable and versatile enough to fit for unique testing needs.
  • Simple to operate and maintain
  • Consistently accurate and reliable measurements
  • Configurable to each users unique needs
  • Meet IP54 specifications for protection from water
These meters are simple to operate and offer an intuitive user interface allowing for easy set up and use from day one. Step-by-step on screen prompts enable both new and seasoned users to operate the meter easily and effectively, resulting in consistently accurate measurements.
Certifications: Calibrated pH meters and kits include a Troemner ISO/IEC accredited calibration certificate traceable to NIST. The ISO/IEC 17025 accredited calibration certificate includes electrical simulation of multiple data points within the functional ranges with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
订购须知 Stand is not included with the meter kits. Each benchtop meter can be configured to individual needs with the variety of optional electrodes, stands with stirrers, and accessories available.