Surface active cleaning agent, decon® 90
供应商: Decon Laboratories UK
DECND905EA 550 CNY
DECND905 DECND901 DECND9020
Surface active cleaning agent, decon® 90
清洗剂 洗涤剂和表面清洁剂
A surface active cleaning agent and/or radioactive decontaminant, for laboratory, medical and specialised industrial applications. A concentrate that is suitable for ‘manual’ cleaning, or for use in an ultrasonic cleaning bath.
  • Biodegradable, phosphate-free and enzyme-free
  • Bactericidal
  • Highly alkaline >pH 13
Certifications: decon® 90 satisfies the criteria in BS EN 1040 1997 for bacterial activity.
Caution: Not suitable for domestic use nor suitable for use on non-ferrous metals, notably aluminium and zinc, or on polycarbonate.