ESD safety shoes, lace-up, uvex business casual, 9512
供应商: Uvex
UVEX9512.2/39EA 1425 CNY
UVEX9512.2/39 UVEX9512.2/40 UVEX9512.2/41 UVEX9512.2/42 UVEX9512.2/43 UVEX9512.2/44 UVEX9512.2/45 UVEX9512.2/46 UVEX9512.2/47 UVEX9512.2/48 UVEX9512.2/38
ESD safety shoes, lace-up, uvex business casual, 9512
Lightweight, casual business shoes made from breathable leather upper, soft padding on collar and tongue plus a self-cleaning, lightweight DUO PU sole resistant to oil and petrol, short-term heat resistance up to 120 °C.
  • Climazone technology for improved comfort
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Wingtip Derby brogue
  • Slip and abrasion resistant with a shock-absorbent sole
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Colour: Black
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S1P, SRC