琼脂糖

供应商: BIO RAD

BRAD162-0100EA 0 CNY
BRAD162-0100
琼脂糖
琼脂糖
溶点: 60…90 °C
存储条件: Ambient
MDL编号: MFCD00081294
CAS号: 9012-36-6
EINECS: 232-731-8
https://cn.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR