Liquid viewer, Apollo II
供应商: Adelphi Manufacturing
MSPP6031511EA 0 CNY
MSPP6031511
Liquid viewer, Apollo II
Designed for use in pharmaceutical and laboratory environments, the Apollo II Liquid Viewer provides a simple method for the detection of visible particles in ampoules, vials, and bottles.
  • Adjustable lamp unit for comfortable working height
  • Uses two low energy LED lamps
  • Fold up frame for space saving and easy storage
  • Removable diffuser for use with coloured glass
  • Intuitive assembly with no tools required
The unit is operated by simply swirling each individual container in front of both panels, and observing for any particulate contamination (other than gas bubbles) unintentionally present in the solution. In front of the black panel, light coloured particulates will be visible, and in front of the white panel, dark coloured particulates will be visible.
The unit uses 2×10 W LED Bulbs (550 mm long) with a diffuser to give light level of between 2000 and 3750 Lux. The bulbs have a service life of 50 000 hours.
W×D×H: 616×417×445 mm
Weight: 7,9 kg
Certifications: Conforms to the European Pharmacopoeia Specification 2.9.20 and the US Pharmacopoeia Specification chapter 41; section 6.1. Adelphi Manufacturing’s ISO 9001:2015 certification guarantees the high quality finish of this product.
订购须知 Supporting IQ/OQ documentation can be supplied. User manuals are available in English, Spanish, French, German, Dutch and Czech – additional languages are also available upon request.
Packaging: Eco-friendly packaging – the product is packaged in sustainably sourced foam, made from polyethylene resin which is derived from renewable sugarcane-based feedstock.